ContactUsLinkComponent
9 components: 2mg/l each of Bromides (Br-) Bromates (BrO3-) Chlorates (ClO3-) Chlorides (Cl-) Fluorides (F-) Nitrites as N (N (NO2-)) Nitrates as N (N (NO3-)) Phosphates as P (P (PO43-)) Sulphates (SO42-) in Water ISO 17025 Conservati 1 * 2 Bottle
Katalogové číslo CMPLRM060731L1.2
New Product
9 components: 2mg/l each of Bromides (Br-) Bromates (BrO3-) Chlorates (ClO3-) Chlorides (Cl-) Fluorides (F-) Nitrites as N (N (NO2-)) Nitrates as N (N (NO3-)) Phosphates as P (P (PO43-)) Sulphates (SO42-) in Water ISO 17025 Conservati 1 * 2 Bottle
Katalogové číslo CMPLRM060731L1.2