ContactUsLinkComponent
MFT Pro 264 PU palm coated, ultrathin seamless polyester microfiber/nylon liner for general purpose use. Excellent breathebility. 1 * 12 PAIR
Catalog # BESTMFTPRO264-8/L
Supplier:  Showa
New Product
MFT Pro 264 PU palm coated, ultrathin seamless polyester microfiber/nylon liner for general purpose use. Excellent breathebility. 1 * 12 PAIR
Catalog # BESTMFTPRO264-8/L
Supplier:  Showa