ContactUsLinkComponent
60033 uvex athletic lite dry uvex athletic lite dry * The uvex athletic lite dry is certified in line with Oeko-Tex® Standard 100 and is a particularly lightweight and sensitive safety glove intended for use during mechanical precision work. The matt 1 * 1 items
Catalog # UVEX60033
Supplier:  UVEX
New Product
60033 uvex athletic lite dry uvex athletic lite dry * The uvex athletic lite dry is certified in line with Oeko-Tex® Standard 100 and is a particularly lightweight and sensitive safety glove intended for use during mechanical precision work. The matt 1 * 1 items
Catalog # UVEX60033
Supplier:  UVEX