Side shelf for NCVI2424 antivibration floor model table, 24 in., load capacity 120 lbs 1 * 1 items
Catalog # ECHLNLSS2424
Dodavatel: DISCOVER ECHO
New Product
Side shelf for NCVI2424 antivibration floor model table, 24 in., load capacity 120 lbs 1 * 1 items
Catalog # ECHLNLSS2424
Dodavatel: DISCOVER ECHO
New Product
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Sealing film for manual application, Parafilm® M
Dodavatel: HEATHROW
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Dodavatel: VWR Collection
VWR®, Transfer Pipettes, High Performance
Dodavatel: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Dodavatel: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Dodavatel: VWR Collection
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR® Facial Tissues
Dodavatel: VWR Collection