ContactUsLinkComponent
,,26 components: 100ug/ml each of Ag Al As B Ba Bi Ca Cd Co Cr Cu Fe Ga K Li Mg Mn Na Ni Pb Sb Sn Sr Ti V Zn in HNO3 8% HF 1% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store 1 * 1 L
Catalog # CMPLRM1115491L
New Product
,,26 components: 100ug/ml each of Ag Al As B Ba Bi Ca Cd Co Cr Cu Fe Ga K Li Mg Mn Na Ni Pb Sb Sn Sr Ti V Zn in HNO3 8% HF 1% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store 1 * 1 L
Catalog # CMPLRM1115491L