PEPMAP NEO C18 5UM 300UMX5MM TRAP 1500B 1 * 3 items
Catalog # HYPE174500
Supplier: Thermo Fisher Scientific
New Product
PEPMAP NEO C18 5UM 300UMX5MM TRAP 1500B 1 * 3 items
Catalog # HYPE174500
Supplier: Thermo Fisher Scientific
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Filter plates, 96-well, for protein purification, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
BOVINE SERUM ALBUMIN (BSA) 20MG/ML 1 * 1 KIT
Supplier: Thermo Fisher Scientific