30 ml 6 round well sterile PS cell culture plate. Barcoded with lid and sterile. (Case of 5) 1 * 5 items
Catalog # 732-4429
Supplier: VWR Collection
New Product
30 ml 6 round well sterile PS cell culture plate. Barcoded with lid and sterile. (Case of 5) 1 * 5 items
Catalog # 732-4429
Supplier: VWR Collection
New Product
Frequently Bought Together
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
VWR® Autoclavable Bags, Polypropylene, Clear
Supplier: VWR Collection
Glass screw top 0,3 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Clear
Supplier: DWK Life Sciences
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Nitric acid 0.2 mol/l (0.2 N)
Supplier: VWR Collection
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals