ContactUsLinkComponent
12 components: 50mg/l each of Tris(2.3-dibromopropyl)phosphate [CAS:126-72-7] Tri-o-cresylphosphate (TOCP) [CAS:78-30-8] Tris(2-chloroethyl)phosphate [CAS:115-96-8] Triphenylphosphate [CAS:115-86-6] Tris(2-chloroisopropyl)phosphate [CAS:13674-8 1 * 1 mL
Catalog # CMPLRD1172191.5
New Product
12 components: 50mg/l each of Tris(2.3-dibromopropyl)phosphate [CAS:126-72-7] Tri-o-cresylphosphate (TOCP) [CAS:78-30-8] Tris(2-chloroethyl)phosphate [CAS:115-96-8] Triphenylphosphate [CAS:115-86-6] Tris(2-chloroisopropyl)phosphate [CAS:13674-8 1 * 1 mL
Catalog # CMPLRD1172191.5