ContactUsLinkComponent
Boron Trifluoride, 14 Percent in Methanol is used as a reagent in organic chemistry most often as a Lewis acid. It is known to be a versatile building block in several other boron compounds. Ungraded products supplied by Spectrum are indicative of a 1 * 1 items
Catalog # SPCMB2644-100ML
Supplier:  Spectrum Chemical
New Product
Boron Trifluoride, 14 Percent in Methanol is used as a reagent in organic chemistry most often as a Lewis acid. It is known to be a versatile building block in several other boron compounds. Ungraded products supplied by Spectrum are indicative of a 1 * 1 items
Catalog # SPCMB2644-100ML
Supplier:  Spectrum Chemical