ContactUsLinkComponent
Twin-tank water softener PG 8597 for continuous provision of soft water with water hardness levels of up to 40 °dH 1 * 1 items
Catalog # MIEL12687200
Supplier:  MIELE
New Product
Twin-tank water softener PG 8597 for continuous provision of soft water with water hardness levels of up to 40 °dH 1 * 1 items
Catalog # MIEL12687200
Supplier:  MIELE