ContactUsLinkComponent
ICP Calibration Standard - Quality Control (XVI) 21 components: 100mg/l each of As Be Ca Cd Co Cr Cu Fe Li Mg Mn Mo Ni Pb Sb Se Sr Ti Tl V Zn in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 36 months. Stor 1 * 100 mL
Catalog # CMPL109487.L1
New Product
ICP Calibration Standard - Quality Control (XVI) 21 components: 100mg/l each of As Be Ca Cd Co Cr Cu Fe Li Mg Mn Mo Ni Pb Sb Se Sr Ti Tl V Zn in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 36 months. Stor 1 * 100 mL
Catalog # CMPL109487.L1