Barium chloride solution for 1000 ml, c(BaCl2) = 0.05 mol/l (0.1 N) Titrisol 1 * 1 Ampoul
Catalog # 1099620001.
Supplier: Merck
New Product
Barium chloride solution for 1000 ml, c(BaCl2) = 0.05 mol/l (0.1 N) Titrisol 1 * 1 Ampoul
Catalog # 1099620001.
Supplier: Merck
New Product
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Weighing boats, PYREX®
Supplier: SciLabware
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific