Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SILIBEADS TYP S 0.60 - 0.75 MM 1 * 10 kg
Catalog # LINR4502-S1-10KG
Supplier:
SIGMUND LINDNER
New Product
Expand image
SILIBEADS TYP S 0.60 - 0.75 MM 1 * 10 kg
Catalog # LINR4502-S1-10KG
Supplier:
SIGMUND LINDNER
New Product
Frequently Bought Together
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Funnels with Short Stem
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
VWR®, Beakers, Tall Form
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Beakers, Low Form
Supplier: VWR Collection