ContactUsLinkComponent
40 components: 1mg/l each of Al Ag As B Ba Be Bi Ca Cd Co Cs Cr Cu Fe Ga Ge In K Li Mg Mn Mo Na Ni Nb Pb Pd Rb Sb Se Sn Sr Ta Ti Tl U V W Zn Zr in HNO3 2% Conservation 12 m 1 * 500 mL
Catalog # CMPLRM100322L5
New Product
40 components: 1mg/l each of Al Ag As B Ba Be Bi Ca Cd Co Cs Cr Cu Fe Ga Ge In K Li Mg Mn Mo Na Ni Nb Pb Pd Rb Sb Se Sn Sr Ta Ti Tl U V W Zn Zr in HNO3 2% Conservation 12 m 1 * 500 mL
Catalog # CMPLRM100322L5