POLY TEMP 4 INNER DOOR TSX60086V 1 * 1 items
Catalog # REVCTSX4IDPTS600
Supplier: REVCO TECHNOLOGIES
New Product
POLY TEMP 4 INNER DOOR TSX60086V 1 * 1 items
Catalog # REVCTSX4IDPTS600
Supplier: REVCO TECHNOLOGIES
New Product
Frequently Bought Together
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Reagent Reservoirs
Supplier: VWR Collection
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Blue Nitrile™
Supplier: KIMBERLY CLARK