ContactUsLinkComponent
Stainless steel slide-in drip tray, 15 mm rim (may affect the temperature distribution) cannot be used in connection with option K 1 MAX. LOADING 8 KG 1 * 1 items
Catalog # MEMMB02075UF750TS
Supplier:  MEMMERT
New Product
Stainless steel slide-in drip tray, 15 mm rim (may affect the temperature distribution) cannot be used in connection with option K 1 MAX. LOADING 8 KG 1 * 1 items
Catalog # MEMMB02075UF750TS
Supplier:  MEMMERT