ContactUsLinkComponent
Stainless steel slide-in drip tray, 15 mm rim (may affect the temperature distribution) - cannot be used in connection with option K 1 1 * 1 items
Catalog # MEMMB02075IPP750EC
Supplier:  MEMMERT
New Product
Stainless steel slide-in drip tray, 15 mm rim (may affect the temperature distribution) - cannot be used in connection with option K 1 1 * 1 items
Catalog # MEMMB02075IPP750EC
Supplier:  MEMMERT