ContactUsLinkComponent
Stainless steel tray, non-perforated, 15 mm rim, may affect the temperature distribution, for ipp 1 * 1 items
Catalog # MEMMB02072IPP55
Supplier:  MEMMERT
New Product
Stainless steel tray, non-perforated, 15 mm rim, may affect the temperature distribution, for ipp 1 * 1 items
Catalog # MEMMB02072IPP55
Supplier:  MEMMERT