ContactUsLinkComponent
10 components: al 2mg/l ca 50mg/l fe 2mg/l k 20mg/l li 1mg/l mg 50mg/l mn 2mg/l na 50mg/l p 10mg/l sr 1mg/l in hno3 0.065% crm, iso 17034 and iso 17025 1 * 500 mL
Catalog # ACSD60F5.2.0065NL5
Supplier:  CPACHEM
New Product
10 components: al 2mg/l ca 50mg/l fe 2mg/l k 20mg/l li 1mg/l mg 50mg/l mn 2mg/l na 50mg/l p 10mg/l sr 1mg/l in hno3 0.065% crm, iso 17034 and iso 17025 1 * 500 mL
Catalog # ACSD60F5.2.0065NL5
Supplier:  CPACHEM