ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 7x4=28 plates, 35mm high , stainless steel, dimensions (HxDxW) 260x360x13 5mm 1 * 1 items
Catalog # TENATE35302
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 7x4=28 plates, 35mm high , stainless steel, dimensions (HxDxW) 260x360x13 5mm 1 * 1 items
Catalog # TENATE35302
Supplier:  TENAK