ContactUsLinkComponent
Standard rack for upright freezers for (HxD) 7x4=28 cryoboxes, 50mm high, stainless steel, dimensions (HxDxW) 389x561x139mm, deliver ed including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23138B10
Supplier:  TENAK
New Product
Standard rack for upright freezers for (HxD) 7x4=28 cryoboxes, 50mm high, stainless steel, dimensions (HxDxW) 389x561x139mm, deliver ed including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23138B10
Supplier:  TENAK