ContactUsLinkComponent
Standard rack for upright freezers for (HxD) 10x4=40 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 361x561x139mm, delive red including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23070B10
Supplier:  TENAK
New Product
Standard rack for upright freezers for (HxD) 10x4=40 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 361x561x139mm, delive red including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23070B10
Supplier:  TENAK