ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 4x3=12 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 320x425x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11266B10
Supplier:  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 4x3=12 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 320x425x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11266B10
Supplier:  TENAK