Eco AluRack for upright freezers for (HxD) 4x3=12 cryoboxes, 40mm high, aluminium, dimensions (HxDxW) 171x425x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Katalogové číslo TENATE11010B10
Dodavatel: TENAK