ContactUsLinkComponent
ICE MACHINE AF 87 AS OX AF 87 AS OX - 68kg - XSafe Self Contained Flake Ice Maker Dimensions : W: 535mm D: 620mm H: 890mm 68kg daily production rate 25kg storage bin capacity Residual water content 25% Air cooled version XSafe - Natural Sanitation Sy 1 * 1 items
Catalog # SCOMAF87ASOX
Supplier:  SCOTSMAN
New Product
ICE MACHINE AF 87 AS OX AF 87 AS OX - 68kg - XSafe Self Contained Flake Ice Maker Dimensions : W: 535mm D: 620mm H: 890mm 68kg daily production rate 25kg storage bin capacity Residual water content 25% Air cooled version XSafe - Natural Sanitation Sy 1 * 1 items
Catalog # SCOMAF87ASOX
Supplier:  SCOTSMAN