ContactUsLinkComponent
Non-slip, flat rubber mat to be placed on the integrated shaker’s platform and optionally on the perforated platform 2000, e.g. for the multi-tier attachment 2000. Material: EPDM Characteristics: high resistance to temperatures up to 134 °C, acids an 1 * 1 items
Catalog # HEID23-03-08-02-66
New Product
Non-slip, flat rubber mat to be placed on the integrated shaker’s platform and optionally on the perforated platform 2000, e.g. for the multi-tier attachment 2000. Material: EPDM Characteristics: high resistance to temperatures up to 134 °C, acids an 1 * 1 items
Catalog # HEID23-03-08-02-66