ContactUsLinkComponent
,,ICP-MS Precious Metals Standard - 8 components: 10mg/l each of Au Ir Os Pd Pt Re Rh Ru in HCl 5% 1 * 100 mL
Catalog # ACSDA2E5.10.5C.L1
Supplier:  CPACHEM
New Product
,,ICP-MS Precious Metals Standard - 8 components: 10mg/l each of Au Ir Os Pd Pt Re Rh Ru in HCl 5% 1 * 100 mL
Catalog # ACSDA2E5.10.5C.L1
Supplier:  CPACHEM