ContactUsLinkComponent
3 components: 217.475mg/l each of n-Tetradecane [CAS:629-59-4] n-Pentadecane [CAS:629-62-9] n-Hexadecane [CAS:544-76-3] in n-Hexane CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2C to 8C 1 * 3 Ampoul
Catalog # CMPLRD0772781.3
New Product
3 components: 217.475mg/l each of n-Tetradecane [CAS:629-59-4] n-Pentadecane [CAS:629-62-9] n-Hexadecane [CAS:544-76-3] in n-Hexane CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2C to 8C 1 * 3 Ampoul
Catalog # CMPLRD0772781.3