ContactUsLinkComponent
1 component: N (NO2-) 0.150mg/l in H2O CRM, ISO 17034 and ISO 17025,,,, Conservation 8 months.,,,, Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
Catalog # CMPLRM051707L1
New Product
1 component: N (NO2-) 0.150mg/l in H2O CRM, ISO 17034 and ISO 17025,,,, Conservation 8 months.,,,, Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
Catalog # CMPLRM051707L1