ContactUsLinkComponent
Sontara micropure mop white 300x590mm, cleanroom & electronics nonwoven wipe (recommended to protect iso 7-9 cleanrooms from particles and additives): strong, durable, absorbent, low-linting, 4 packs x 500 wipes per box. 1 * 500 items
Catalog # SONT1000001898
Supplier:  SONTARA
New Product
Sontara micropure mop white 300x590mm, cleanroom & electronics nonwoven wipe (recommended to protect iso 7-9 cleanrooms from particles and additives): strong, durable, absorbent, low-linting, 4 packs x 500 wipes per box. 1 * 500 items
Catalog # SONT1000001898
Supplier:  SONTARA