sontara micropure 100 white 224x224mm, cleanroom & electronics nonwoven wipe (recommended for iso 5-8* cleanrooms): strong, durable, absorbent, low-linting. 1 * 300 items
Catalog # SONT1000001885_U
Supplier: SONTARA
New Product
sontara micropure 100 white 224x224mm, cleanroom & electronics nonwoven wipe (recommended for iso 5-8* cleanrooms): strong, durable, absorbent, low-linting. 1 * 300 items
Catalog # SONT1000001885_U
Supplier: SONTARA
New Product
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Small head swabs, Alpha®
Supplier: Texwipe
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Cleanroom wipes, Amplitude™ Theta™
Supplier: CONTEC