BCA Protein Assay Kit with Dilution-Free BSA Protein Standard Concentration - BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL Kit Contents - 2 x 1 mL 8-channel tubestrip, prediluted Quantity - 1 Kit (500 mL) 1 * 1 KIT
Catalog # PIERA55865
Dodavatel: Thermo Fisher Scientific