ContactUsLinkComponent
Vivaflow SU is a next generation plug-and-play tangential flow filtration (TFF) cassette for ultrafiltration and diafiltration of in termediate sample volumes. It has been specifically designed for ease of use in research and development laboratories 1 * 1 Each
Catalog # SARTVF-S050H0100IV
Supplier:  Sartorius
New Product
undefined
Vivaflow SU is a next generation plug-and-play tangential flow filtration (TFF) cassette for ultrafiltration and diafiltration of in termediate sample volumes. It has been specifically designed for ease of use in research and development laboratories 1 * 1 Each
Catalog # SARTVF-S050H0100IV
Supplier:  Sartorius