ContactUsLinkComponent
9 Panel Urine Round Cup (AMP1000/ BZO300 /COC300 /MDMA500 /MTD300 /OPI2000 /PPX300 /THC50 /K250 +A&B ADLT) null
Catalog # SURERCUP9DS1AB
Supplier:  SURESCREEN
New Product
9 Panel Urine Round Cup (AMP1000/ BZO300 /COC300 /MDMA500 /MTD300 /OPI2000 /PPX300 /THC50 /K250 +A&B ADLT) null
Catalog # SURERCUP9DS1AB
Supplier:  SURESCREEN