EU Drinking Water Directive PFAS Mixture 51 10 µg/mL in Methanol:Water 1 * 1 mL
Catalog # EHERA30000051MW
Supplier: EHRENSTORFER
New Product
EU Drinking Water Directive PFAS Mixture 51 10 µg/mL in Methanol:Water 1 * 1 mL
Catalog # EHERA30000051MW
Supplier: EHRENSTORFER
New Product
Frequently Bought Together
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
VWR® Reagent Reservoirs
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid) ≥99.5%, Ultrapure
Supplier: Apollo Scientific