Custom repliQa HiFi ToughMix including GelTrack Loading Dye,25mL 1 * 1 KIT
Catalog # QUNT172-5599
Supplier: Quantabio
New Product
Custom repliQa HiFi ToughMix including GelTrack Loading Dye,25mL 1 * 1 KIT
Catalog # QUNT172-5599
Supplier: Quantabio
New Product
Frequently Bought Together
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
PMAxx™ dye 20 mM in water
Supplier: Biotium
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals