,,22 components: 10mg/l each of Be Cd As Cu Co Mn Pb Tl V Cr Sb Sn Zn Ag Ba Mo Te W Se Hg Li Mg in HNO3 2% , ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM108688L1
Supplier: CUSTOM MADE CHEMICALS LAB