ContactUsLinkComponent
,,10 components: Ethanol [CAS:64-17-5] 50mg/kg Acetone [CAS:67-64-1] 30mg/kg Methyl-tert.butylether [CAS:1634-04-4] 10mg/kg 2-Butanol [CAS:78-92-2] 10mg/kg 2-Propanol [CAS:67-63-0] 10mg/kg Toluene [CAS:108-88-3] 10mg/kg Benzene [CAS:71-43 1 * 10 mL
Catalog # CMPLRD11498110
New Product
,,10 components: Ethanol [CAS:64-17-5] 50mg/kg Acetone [CAS:67-64-1] 30mg/kg Methyl-tert.butylether [CAS:1634-04-4] 10mg/kg 2-Butanol [CAS:78-92-2] 10mg/kg 2-Propanol [CAS:67-63-0] 10mg/kg Toluene [CAS:108-88-3] 10mg/kg Benzene [CAS:71-43 1 * 10 mL
Catalog # CMPLRD11498110