ContactUsLinkComponent
,,21 components: 100mg/l each of 2-Chlorophenol [CAS:95-57-8] 3-Chlorophenol [CAS:108-43-0] 4-Chlorophenol [CAS:106-48-9] 2 ,3-Dichlorophenol [CAS:576-24-9] 2,4-Dichlorophenol [CAS:120-83-2] 2 ,5-Dichlorophenol [CAS:583-78-8] 2,6-Dichloro 1 * 1 Ampoul
Catalog # CMPLRD0467541.2
New Product
,,21 components: 100mg/l each of 2-Chlorophenol [CAS:95-57-8] 3-Chlorophenol [CAS:108-43-0] 4-Chlorophenol [CAS:106-48-9] 2 ,3-Dichlorophenol [CAS:576-24-9] 2,4-Dichlorophenol [CAS:120-83-2] 2 ,5-Dichlorophenol [CAS:583-78-8] 2,6-Dichloro 1 * 1 Ampoul
Catalog # CMPLRD0467541.2