ContactUsLinkComponent
SkillPak Super A 5mL col. Column dimension 0.8 cm ID × 10 cm bed height Standard flow rate: 1.3 mL/min (150 cm/h) Maximum flow rate: 5 mL/min (600 cm/h) for TOYOPEARL M and C grade resins 2.5 mL/min (300 cm/h) for TSKgel, Ca++Pure-HA and TOYOPEARL S 1 * 1 items
Catalog # TOYO0045400
Supplier:  TOSOH Bioscience
New Product
SkillPak Super A 5mL col. Column dimension 0.8 cm ID × 10 cm bed height Standard flow rate: 1.3 mL/min (150 cm/h) Maximum flow rate: 5 mL/min (600 cm/h) for TOYOPEARL M and C grade resins 2.5 mL/min (300 cm/h) for TSKgel, Ca++Pure-HA and TOYOPEARL S 1 * 1 items
Catalog # TOYO0045400
Supplier:  TOSOH Bioscience