ContactUsLinkComponent
MELTING POINT STANDARD PHENACETIN 134 C TO 136 C 1 * 1 items
Catalog # REGERMP132
Supplier:  REAGECON
New Product
MELTING POINT STANDARD PHENACETIN 134 C TO 136 C 1 * 1 items
Catalog # REGERMP132
Supplier:  REAGECON