POTASSIUM BROMATE KBR/KBRO3 0.5 MOL/L 1 * 1 L
Catalog # CMPLZ14424556-1L
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
POTASSIUM BROMATE KBR/KBRO3 0.5 MOL/L 1 * 1 L
Catalog # CMPLZ14424556-1L
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
pH indicator papers, universal/special
Supplier: MACHEREY-NAGEL
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, chloride, Spectroquant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach