AMMONIA SOLUTION 25-30% VLSI 1 * 180 kg
HONC17326-180KG
New Product
Frequently Bought Together
2-Propanol USP, J.T.Baker®
: Avantor
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
: Thermo Fisher Scientific
4-Methyl-2-pentanone 99.5% for analysis
: Thermo Fisher Scientific
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
4-Methyl-2-pentanone ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
4-(Methylamino)phenol sulphate 99%
: Thermo Fisher Scientific
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
1,2-Dichloroethane ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
n-Heptane 99+%, extra pure
: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
: Honeywell Chemicals
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific