ContactUsLinkComponent
8 components: 100mg/l each of Al Cr Cu Fe Mn Ba Zn Zr in HNO3 5% 1 * 100 mL
Catalog # CMPLRM113431L1
New Product
8 components: 100mg/l each of Al Cr Cu Fe Mn Ba Zn Zr in HNO3 5% 1 * 100 mL
Catalog # CMPLRM113431L1