ContactUsLinkComponent
20mm combination seal : Aluminium capp, gold lacquered, complete tear off, moulded septa butyl, dark grey 55 shore A 3,0mm 1 * 1.000 items
Catalog # LAPH20031532
Supplier:  LA PHA PACK
New Product
20mm combination seal : Aluminium capp, gold lacquered, complete tear off, moulded septa butyl, dark grey 55 shore A 3,0mm 1 * 1.000 items
Catalog # LAPH20031532
Supplier:  LA PHA PACK