ContactUsLinkComponent
The performance option increases the number of installed Peltier modules in size 750 from 2 to 4 units. This enhances performance and improves technical specifications. 1 * 1 items
Catalog # MEMMHPP750ECOP1
Supplier:  MEMMERT
New Product
The performance option increases the number of installed Peltier modules in size 750 from 2 to 4 units. This enhances performance and improves technical specifications. 1 * 1 items
Catalog # MEMMHPP750ECOP1
Supplier:  MEMMERT