ContactUsLinkComponent
8 components: Calcium (Ca) 200mg/l Chlorides (Cl-) 750mg/l Phosphorus (P) 5mg/l Sulphur (S) 100mg/l Magnesium (Mg) 50mg/l Sodium (Na) 100mg/l Potassium (K) 50mg/l Carbon (C) 50mg/l in Nitric Acid 5% Hydrofluoric acid 0.1% ISO 17034 an 1 * 500 mL
Catalog # CMPL8B22.50.L5.2
New Product
8 components: Calcium (Ca) 200mg/l Chlorides (Cl-) 750mg/l Phosphorus (P) 5mg/l Sulphur (S) 100mg/l Magnesium (Mg) 50mg/l Sodium (Na) 100mg/l Potassium (K) 50mg/l Carbon (C) 50mg/l in Nitric Acid 5% Hydrofluoric acid 0.1% ISO 17034 an 1 * 500 mL
Catalog # CMPL8B22.50.L5.2