CLEARVUE MOUNTANT XYL 1 * 2 items
Catalog # SHAN4212
Supplier: SHANDON
New Product
Frequently Bought Together
Total Iron Test Strips, Dosatest, VWR®
Supplier: VWR Chemicals
EOSIN RAL 555 1L 1 * 1 L
Supplier: RAL
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
VWR® Kjeldahl Tablets
Supplier: VWR Chemicals
Test strips, water hardness, AQUADUR®
Supplier: MACHEREY-NAGEL
Test kits, phosphate (kits and reagents) CHECKIT®
Supplier: Lovibond
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Supplier: Honeywell Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Sodium hydroxide 98.5%, pellets for analysis
Supplier: Thermo Fisher Scientific