BLUING REAGENT 3800 ML 1 * 1 items
Catalog # SHAN7301
Supplier: SHANDON
New Product
Frequently Bought Together
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Powder pillows, sulphate, Sulfa 4
Supplier: Lovibond
BOND EPITOPE RETRIEVAL SOLUTION 2 1 * 1 L
Supplier: LEICA MICROSYSTEMS
Colorimetric rapid tests, sulphide, VISOCOLOR® ECO
Supplier: MACHEREY-NAGEL
Reagents for ammonia determination using Lovibond® MD photometers
Supplier: Lovibond
SODIUM METHOXIDE 0.1M 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
PARAPLAST REGULAR BULK PACK 56°C 1 * 5 kg
Supplier: LEICA HISTOLOGY
pH-Indicator, Litmus paper
Supplier: Lach-ner S.R.O.
Sodium Hydroxide solution 32 % w/w for nitrogen determination 1 * 25 L
Supplier: PanReac AppliChem