Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
NEG-50 MULTI-PACK 6x118ml 1 * 4 items
Catalog # SHAN6506M
Supplier:
SHANDON
New Product
Expand image
NEG-50 MULTI-PACK 6x118ml 1 * 4 items
Catalog # SHAN6506M
Supplier:
SHANDON
New Product
Frequently Bought Together
pH Papers, without color scale
Supplier: MACHEREY-NAGEL
50 TABLETS ORGAPHOS-OX PHOTOMETER 1 * 50 items
Supplier: WATER ID PRIMELAB
VARIO SG REAGENT SET 1 * 1 SET
Supplier: Lovibond
Nitrite VHR L, 500 ml
Supplier: Lovibond
TISSUE MARKING DYE ORANGE,59ML 1 * 1 items
Supplier: SHANDON